Learn More
Synthware™ Solid Phase Peptide Synthesis Vessel, Ground Glass Joint, Side Arm
Solid Phase Peptide Synthesis Vessel with fritted glass disc and a lower ground glass joint and side connection.
Supplier: Synthware™ P140025M
Chemical Identifiers
| Organic Synthesis | |
| Borosilicate Glass | |
| Vessel with T-bore PTFE stopcock and ground joint |
Specifications
| Borosilicate Glass | |
| Vessel with T-bore PTFE stopcock and ground joint | |
| 25 mL | |
| Organic Synthesis |