Learn More
Synthware™ Solid Phase Peptide Synthesis Vessel, Ground Glass Joint, Side Arm
GREENER_CHOICE
Catalog No. 31501579
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
31501579 10 mL
31501580 25 mL
31501581 -
31501582 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 31501579 Supplier Synthware™ Supplier No. P140010M
Solid Phase Peptide Synthesis Vessel with fritted glass disc and a lower ground glass joint and side connection.
Specifications
For Use With (Application) Organic Synthesis
Product Type Vessel with T-bore PTFE stopcock and ground joint
Material Borosilicate Glass
Capacity (Metric) 10 mL