Learn More
Eisco™ Solid Neoprene Stoppers
Pack of ten solid neoprene stoppers. Made from long lasting neoprene rubber. Oil resistant, use up to 250°F / 120°C. Suitable for use with petroleum products, minerals, oils, fats as well as most inorganic acids.
$6.13 - $192.67
Specifications
| Description | Pack of 10 |
|---|---|
| Autoclavable | Autoclavable |
| Color | Blue |
| Material | Neoprene |
| Product Type | Steri-stopper |
| Catalog Number | Mfr. No. | Diameter (Metric) Bottom | Diameter (Metric) | Height (Metric) | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) Bottom | Diameter (Metric) | Height (Metric) | Price | Quantity | |||
|
S40971
|
Eisco™
CH0319ANUP |
6 mm | 8 mm | 16 mm |
Pack of 10 Each for $6.13
|
|
|||
|
S40979
|
Eisco™
CH0319BNUP |
8 mm | 10.5 mm | 20 mm |
Pack of 10 Each for $7.63
|
|
|||
|
S40985
|
Eisco™
CH0319DNUP |
10 mm | 12.5 mm | 20 mm |
Pack of 10 Each for $9.47
|
|
|||
|
S40987
|
Eisco™
CH0319ENUP |
11 mm | 14 mm | 24 mm |
Pack of 10 Each for $12.23
|
|
|||
|
S40989
|
Eisco™
CH0319FNUP |
13 mm | 16 mm | 24 mm |
Pack of 10 Each for $15.33
|
|
|||
|
S40996
|
Eisco™
CH0319GNUP |
15 mm | 18 mm | 24 mm |
Pack of 10 Each for $19.53
|
|
|||
|
S41004
|
Eisco™
CH0319HNUP |
17 mm | 20 mm | 26 mm |
Pack of 10 Each for $25.17
|
|
|||
|
S41007
|
Eisco™
CH0319INUP |
18 mm | 21 mm | 26 mm |
Pack of 10 Each for $30.90
|
|
|||
|
S41010
|
Eisco™
CH0319JNUP |
19 mm | 22 mm | 28 mm |
Pack of 10 Each for $33.53
|
|
|||
|
S41017
|
Eisco™
CH0319KNUP |
21 mm | 24 mm | 28 mm |
Pack of 10 Each for $37.97
|
|
|||
|
S41022
|
Eisco™
CH0319LNUP |
23 mm | 26 mm | 28 mm |
Pack of 10 Each for $47.10
|
|
|||
|
S41026
|
Eisco™
CH0319MNUP |
25 mm | 28 mm | 28 mm |
Pack of 10 Each for $55.37
|
|
|||
|
S41032
|
Eisco™
CH0319NNUP |
27 mm | 31 mm | 32 mm |
Pack of 10 Each for $70.93
|
|
|||
|
S41035
|
Eisco™
CH0319ONUP |
29 mm | 31 mm | 32 mm |
Pack of 10 Each for $74.80
|
|
|||
|
S41038
|
Eisco™
CH0319PNUP |
31 mm | 36 mm | 35 mm |
Pack of 10 Each for $102.50
|
|
|||
|
S41041
|
Eisco™
CH0319QNUP |
33 mm | 38 mm | 38 mm |
Pack of 10 Each for $128.17
|
|
|||
|
S41052
|
Eisco™
CH0319TNUP |
40 mm | 49 mm | 40 mm |
Pack of 10 Each for $192.67
|
|
|||
Description
Warranty
1 year limited manufacturer warranty
Specifications
| Pack of 10 | |
| Blue | |
| Steri-stopper | |
| Non-sterile | |
| Stopper for Labware | |
| 10 |
| Autoclavable | |
| Neoprene | |
| Conical | |
| Reusable | |
| Petroleum products, minerals, oils and fats, as well as most inorganic acids and bases | |
| -40 to 130°C |