missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Solid Glass Penny Head Stoppers
Solid glass penny head stoppers are made from high quality borosilicate glass and come in a varity of joint sizes.
Supplier: Eisco™ CH0870B
Description
Warranty
1 year limited manufacturer warranty
Specifications
| Clear | |
| Labware closure | |
| 1 Ea. | |
| 1.3 cm | |
| Autoclavable | |
| Conical |
| Pennyhead Stopper | |
| 1.3 cm | |
| Borosilicate Glass | |
| 14/23 | |
| 14 mm |