Learn More
Eisco™ Solid Brass Hooked Weights
Solid brass hooked weights with integrated hooks. Weights are cylindrical with natural brass color, weight stamped at top
Supplier:  Eisco™ PHHWBR2
 View more versions of this product
Catalog No. S41592
Only null left
Description
Description
  • Solid brass hooked weights
  • Hook is integrated with weight
  • Natural brass color with stamped identifier at top
  • Weight is cylindrical
Warranty
1 year limited manufacturer warranty
Specifications
Specifications
Cylindrical Weight
20 g
Calibrating scales and balances
1
Brass
5.72 cm
Weight with hook
Product Suggestions
Product Suggestions