Learn More
Thermo Scientific™ Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps.
Supplier:  Thermo Scientific™ SK1830DK
 View more versions of this product
Catalog No. 09034213
Only null left
Description
Description
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps. With exceptional innovation, reliability and accuracy, these accessories are the optimal choice for a busy lab.
Maximize capacity on the Thermo Scientific™ Solaris Orbital Shakers with universal platforms and a variety of clamps to fit most vessels.
Certifications
UL, cUL, CE
Warranty
1 Year
Specifications
Specifications
Dual Stack Universal Platform Upgrade Kit
18 x 30 in.
457.2 mm
762 mm
One platform, screws, supporting rods
1 Year
Certificate of Analysis (COA), Certificate of Conformance (COC)
18 in.
30 in.
Solaris 4000
Solaris
Product Suggestions
Product Suggestions