Learn More
Thermo Scientific™ Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps.
Supplier:  Thermo Scientific™ SK1214D
 View more versions of this product
Catalog No. 09034205
Only null left
Description
Description
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps. With exceptional innovation, reliability and accuracy, these accessories are the optimal choice for a busy lab.
Maximize capacity on the Thermo Scientific™ Solaris Orbital Shakers with universal platforms and a variety of clamps to fit most vessels.
Certifications
UL, cUL, CE
Warranty
1 Year
Specifications
Specifications
Dual Stack Universal Platform
12 x 14 in.
304.8 mm
355.6 mm
Two platforms, screws, and supporting rods
1 Year
Certificate of Analysis (COA), Certificate of Conformance (COC)
12 in.
14 in.
Solaris 2000
Solaris
Product Suggestions
Product Suggestions