missing translation for 'onlineSavingsMsg'
Learn More

ICU Medical SOL-M™ Blood Collection Tube Holder with Luer Lock Adapter

Catalog No. 22024015
Encompass
Change view
Click to view available options
Size:
Standard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22024015 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22024015 Supplier ICU Medical Supplier No. 110201050003

Suitable for use with SOL-CARE™ Safety Blood Collection Needle Infusion Set.

  • Blue tip
  • Includes luer adapter
  • For use with SOL-CARE™ Safety Blood Collection Needle Infusion Set

Specifications

Size Standard
Sterility Sterile
Length (Metric) 5 cm
Product Type Tube Holder
Includes Male Luer Adapter
Length (English) 2 in.
Material Polypropylene
Color Blue Tip
For Use With (Application) Blood Draw