missing translation for 'onlineSavingsMsg'
Learn More

ICU Medical Sol-Care™ Safety Blood Collection Needle with Multi-Sample Luer

Catalog No. 22024008
Encompass
Change view
Click to view available options
Gauge:
21 G
Length (English):
7 in. (tubing)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gauge Length (English)
22024008 21 G 7 in. (tubing)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22024008 Supplier ICU Medical Supplier No. 110201030030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Safety butterfly needle designed to be used for blood collection.

  • Simple safety mechanism that should be activated immediately after blood withdrawal is complete
  • Reduces the risk of needlestick injury
  • Thumb rest to ease handling during safety activation
  • Flexible wings to secure device to patient's skin
  • Thin walled needle is designed to minimize patient discomfort
  • Available in 21, 23 or 25 gauge
  • Transparent housing and tubing for clear visualization of blood flow
  • DEHP-free, flexible, soft, and kink resistant tubing

Specifications

Product Type Butterfly Needle with Luer Adapter
Material PVC
Sterility Sterile
Closure Type Sliding Shield
Color Green
For Use With (Application) Blood Collection
Gauge 21 G
Length (English) 7 in. (tubing)
Length (Metric) 19 mm