missing translation for 'onlineSavingsMsg'
Learn More

LaMotte™ Soil pH TesTabs Kit

Catalog No. S98237
Change view
Click to view available options
Includes:
Laminated color chart, diagrammed instructions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S98237 Laminated color chart, diagrammed instructions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98237 Supplier LaMotte™ Supplier No. LAM5912

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kit for measuring pH.

  • Ideal for younger students and teachers who would like an alternative to liquid pH indicators
  • Simple diagrammed instructions
  • Laminated color chart
  • 50 tests for pH
  • Wide Range: 4 to 11 in 1.0pH units
  • Distilled or deionized water recommended

Specifications

For Use With (Application) For measuring ph
Includes Laminated color chart, diagrammed instructions
Class Size 1 Class
Product Type Soil pH Test Tabs Kit