missing translation for 'onlineSavingsMsg'
Learn More

LaMotte™ Soil Macronutrients Test Kit

Make detailed evaluations of soil composition.

Specifications

Class Size 1 Class
Product Type Soil Macronutrients Test Kit
View More Specs

Includes:
  • Soil sampling tube
  • 50 soil sample bags
  • LaMotte Soil Handbook
  • Downloadable free resource: A Study of Soil Science
  • Soil analysis report forms

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
S17704
View Documents Promotion Details
LaMotte™
LAM 5928
Soil Testing Station N/A
Only null left
 
S17704A
View Documents Promotion Details
LaMotte™
LAM R 5928
Soil Testing Station refill kit N/A
Only null left
 
Description

Description

  • Soil analysis outfit includes all of the materials needed for accurate and detailed soil evaluation
  • Determines soil pH, nitrogen, phosphorus, potassium content and soil texture
  • Each self-contained, individual module contains the necessary materials and equipment to conduct 50 tests
  • All components are contained in a specially designed, rugged carrying case
  • Includes the LaMotte Soil Handbook and downloadable free resource, A Study of Soil Science

Octa Slide Comparator

  • Test Factor: pH (40 tests)
  • Range: 3.8-9.6

Color Chart

  • Test Factor: Nitrogen / Phosphorus (50 tests ea.)
  • Range: 10-150 lb/acre / 10-200 lb/acre

Turbidity Column

  • Test Factor: Potassium (30 tests)
  • Range: 100-400 lb./acre

Order Info

Grades 6+.

Specifications

Specifications

1 Class
Soil Macronutrients Test Kit
Videos
SDS
Documents

Documents