missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOTRAX™ Soil Guard™ Mat
Description
- Specially formulated rubber allows Soil Guard™ to withstand harsh weather environments and heavy foot traffic
- Excels in both indoor and outdoor high traffic entryways and can be hosed off for easy cleaning
- Heavy-duty, durable rubber compound
- Low profile for door clearance / cart traffic
- High-low design scrapes debris from shoe traffic
- Easy to clean – simply hose off
- Recommended and approved product as a part of the GreenTRAX™ program for “Green Cleaning” environments
Specifications
Specifications
| Thickness (English) | 1/4 in. |
| Dimensions (L x W) | 3 x 5 in. |
| Width (English) | 5 in. |
| Quantity | 1 Ea. |
| Color | Black |
| Length (English) | 3 in. |