missing translation for 'onlineSavingsMsg'
Learn More

Moldex™ Softies™ Ecostation™ Ear Plug Dispenser

Catalog No. 19384369
Change view
Click to view available options
CSA Z94.2:
Class AL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. CSA Z94.2
19384369 Class AL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19384369 Supplier Moldex™ Supplier No. MDX6701

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Softies™ EcoStation™ dispensers pre-filled with ear plugs. Easy access, wall-mountable unit. When empty, simply dispose of it and hang a new one.

  • More economical and green than separately packaged ear plugs
  • Easy-to-install sealed refills snap in quickly
  • Empty bottles are fully recyclable
  • Gives workers easy access to hearing protection
  • 500 pairs in each EcoStation
  • High visibility to increase awareness for hearing conservation
  • Hang anywhere ear plugs are needed
  • Workers take one pair at a time with no wasteful grabbing of multiple pairs
  • Independently tested to ANSI S3.19-1974
  • 100% PVC-free

Specifications

ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Construction, Utilities, Manufacturing, Production, Mining, Oil and Gas, and Other Various Industries
High Visibility Yes
Holds 500 pairs
Packaging Type Dispenser
Pairs per Package 500 Pairs/Dispenser
Plug Material Foam
Plug Color Orange
Noise Reduction Rating (NRR) 33 dB
Corded Uncorded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No
Mounting Wall Mountable
Show More Show Less