missing translation for 'onlineSavingsMsg'
Learn More

International Enviroguard™ Soft Scrubs™ Short Sleeve Shirts

Catalog No. 19472451
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19472451 Large
19472447 Small
19472446 Medium
19472448 X-Large
19472443 2X-Large
19472444 3X-Large
19472445 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19472451 Supplier International Enviroguard™ Supplier No. 2055BL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disposable short sleeve SMS shirts are comfortable to wear for long durations and are ideal for controlled environments.

  • Short sleeve SMS shirts offer comfort and safety to work long hours
  • Offer modesty with complete ''no show through'' coverage
  • Can easily be worn as a protective undergarment beneath a sterile gown
  • After use, quickly and easily dispose to reduce spread of germs, bioburdens, and other contagions
  • Round hemmed neck comfortably lays flat
  • Left chest pocket allows storage for eyeglasses, small instruments, and pens
  • Applications include pharmaceuticals, labs, healthcare, controlled environments, and more
  • Anti-static fabric is made of an opaque material that doesn't show through and resists abrasions
  • Convenient, fabric loop for easily hanging up clothing
  • Elastic wrists stretch to provide snug, secure fit

Specifications

Size Large
Gender Unisex
Material SMS
Color Blue
Neck Style Hemmed Crew
Seam Type Serged
Sleeve Style Short Sleeve
Disposable Single-use
Latex Free Yes
No. of Pockets 2
Product Type Scrub Shirt
Sterility Non-sterile
For Use With (Application) Pharmaceutical, Lab, Medical, Manufacturing
Show More Show Less