missing translation for 'onlineSavingsMsg'
Learn More

International Enviroguard™ Soft Scrubs™ Pants

Catalog No. 19472453
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19472453 Small
19472449 Medium
19472436 Large
19472437 X-Large
19472433 2X-Large
19472434 3X-Large
19472435 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19472453 Supplier International Enviroguard™ Supplier No. 2052BS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Soft and comfortable SMS pants feature an elastic waistband for a secure fit. Single-use scrubs offer modesty with complete coverage and can also be worn underneath gowns.

  • Scrub pants offer comfort and safety to work long hours
  • Soft pants offer modesty with complete ''no show through'' coverage
  • Can easily be worn as a protective undergarment beneath a sterile gowns
  • Quickly and easily dispose after each use to reduce spread of germs, bioburdens, and other contagions
  • Elastic waistband offers a snug, secure fit
  • Hip pocket allows storage of eyeglasses, small instruments, pens, and more
  • Possible use areas include pharmaceuticals, labs, healthcare, and controlled environments
  • Anti-static fabric is made of an opaque material that doesn't show through and resists abrasions
  • Convenient fabric loop for easily hanging up clothing
  • Reinforced crotch resists tears

Specifications

Size Small
Gender Unisex
Material SMS
Color Blue
Ankle Style Open
Seam Type Serged
No. of Pockets 1
Disposable Single-use
Latex Free Yes
Sterility Non-sterile
Waist Style Wide Elastic
For Use With (Application) Pharmaceutical, Lab, Healthcare, Controlled Environments
Show More Show Less