missing translation for 'onlineSavingsMsg'
Learn More

International Enviroguard™ Soft Scrubs™ SMS Long Sleeve Shirts

Catalog No. 19472452
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19472452 Large
19472450 Small
19472441 Medium
19472442 X-Large
19472438 2X-Large
19472439 3X-Large
19472440 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19472452 Supplier International Enviroguard™ Supplier No. 2054BL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disposable long sleeve SMS shirts are comfortable to wear for long durations and are ideal for controlled environments.

  • Long sleeve SMS shirts offer comfort and safety to work long hours
  • Offer modesty with complete ''no show through'' coverage
  • Can easily be worn as a protective undergarment beneath a sterile gown
  • After use, quickly and easily dispose to reduce spread of germs, bioburdens, and other contagions
  • Round hemmed neck comfortably lays flat
  • Left chest pocket allows storage for eyeglasses, small instruments, and pens
  • Applications include pharmaceuticals, labs, healthcare, controlled environments, and more
  • Anti-static fabric is made of an opaque material that doesn't show through and resists abrasions
  • Convenient, fabric loop for easily hanging up clothing
  • Elastic wrists stretch to provide snug, secure fit

Specifications

Size Large
Gender Unisex
Material SMS
Color Blue
Neck Style Hemmed Crew
Seam Type Serged
Sleeve Style Long Sleeve
Cuff Style Elastic
Disposable Single-use
Latex Free Yes
No. of Pockets 1
Product Type Scrub Shirt
Sterility Non-sterile
For Use With (Application) Pharmaceutical, Lab, Medical, Manufacturing
Show More Show Less