missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Learning Resources Soft Foam Cross-Section Eye Model
Description
Includes
Activity Guide with lots of facts and a reproducible eye diagram to label.
- One side shows the name of each part; other side is labeled only with letters so kids can quiz themselves
- Features cornea, iris, pupil, retina, lens, optic nerve, vitreous humor, sclera, choroid, macula ciliary muscle, optic disc and zonules
- Measures 5 in. diameter
Order Info
Grades 2+.
Specifications
Specifications
| For Use With (Application) | Open up nature for inner exploration |
| Includes | Parts of the eye like cornea, iris, pupil, retina, lens, optic nerve, vitreous humor, sclera, choroid, macula ciliary muscle, optic disc and zonules |
| Material | Foam |
| Product Type | Model, Eye Cross Section |