missing translation for 'onlineSavingsMsg'
Learn More

Quidel Sofia & Sofia 2 Lyme FIA Test Kit

Catalog No. 23043092
Change view
Click to view available options
Quantity:
25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23043092 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23043092 Supplier Quidel Supplier No. Q20298

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Sofia Lyme Fluorescent Immunoassay (FIA) uses advanced immunofluorescence-based lateral-flow technology to provide rapid differential detection of human IgM and IgG antibodies to Borrelia burgdorferi from serum and plasma specimens from patients suspected of B. burgdorferi infection.

  • Results in 10 minutes - Fast time to results for truly rapid Lyme testing.
  • Self-contained Test Cassette - Clean, easy to use and dispose of.
  • Dual work modes - Adjusts to your volume fluctuations.
  • Differentiated results - IgM and IgG results with a single test.
  • Room temperature storage - No refrigeration space needed. No need to wait for reagents to warm up. Rapid Lyme tests can be run immediately as needed.

Specifications

Content And Storage Room temperature (15°C to 30°C / 59°F to 86°F)
Clia Complexity Moderate
Format Cassette
Sample Type Serum or Plasma
For Use With (Application) Lyme Testing
Includes

Individually Packaged Test Cassettes (25), Borrelia burgdorferi antigens and anti-human IgM/IgG Reagent Vials filled with 0.3 mL reagent solution (25), One (1) Bottle Positive Control: B. burgdorferi IgM and IgG positive plasma diluted 1:10 in 1xPBS with microcide, One (1) Bottle Negative Control: B. burgdorferi negative serum diluted 1:10 in 1xPBS with microcide

Quantity 25
Test Time 10 min.