missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tris(1,1,1,3,3,3-hexafluoroisopropoxy)borohydride 75.0+%, TCI America™

Catalog No. S08105G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08105G Supplier TCI America Supplier No. S08105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 139494-68-1
Molecular Formula C9H3BF18NaO3
Molecular Weight (g/mol) 534.89
MDL Number MFCD16295193
InChI Key FRRJTMMEYJQWIY-UHFFFAOYSA-N
PubChem CID 25112531
IUPAC Name sodium tris[(1,1,1,3,3,3-hexafluoropropan-2-yl)oxy]-λ²-boranuide
SMILES [Na+].FC(F)(F)C(O[B-](OC(C(F)(F)F)C(F)(F)F)OC(C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Color White
Quantity 5 g
UN Number 1409
Formula Weight 535.90
Percent Purity ≥75.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Tris(1,1,1,3,3,3-hexafluoroisopropoxy)borohydride [Reducing Reagent]
TSCA No
Recommended Storage Refrigerator