missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Tripolyphosphate Hexahydrate, Certified Reference Material, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 15091-98-2 |
| Flash Point | Not Applicable |
| Molecular Formula | Na5P3O10 · 6H2O |
| Linear Formula | Na5P3O10 · 6H2O |
| Quantity | 1 g |
| Synonym | Pentasodium tripolyphosphate hexahydrate |
| Molecular Weight (g/mol) | 475.96 |
| Formula Weight | 475.96 |
| Grade | Certified Reference Material |
| Physical Form | Powder |