missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium trifluoroacetate, 98%
Description
Sodium trifluoroacetate is used in acid catalyzed reactions. It is also used as an intermediate for active pharmaceutical ingredients (API). Further, it acts as a precursor for the trifluoromethylation of aldehydes using copper(I) halide as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium trifluoroacetate is used in acid catalyzed reactions. It is also used as an intermediate for active pharmaceutical ingredients (API). Further, it acts as a precursor for the trifluoromethylation of aldehydes using copper(I) halide as a catalyst.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases. Hygroscopic. Moisture sensitive.
Sodium trifluoroacetate is used in acid catalyzed reactions. It is also used as an intermediate for active pharmaceutical ingredients (API). Further, it acts as a precursor for the trifluoromethylation of aldehydes using copper(I) halide as a catalyst.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases. Hygroscopic. Moisture sensitive.
Specifications
Specifications
| Melting Point | ∼207°C (decomposition) |
| Assay Percent Range | 98% |
| Quantity | 100 g |
| Beilstein | 3597949 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water. |
| IUPAC Name | sodium trifluoroacetate |
| Formula Weight | 136 |
| Percent Purity | 98% |
| Chemical Name or Material | Sodium trifluoroacetate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 220-879-6 |
| RTECSNumber | AK0250000 |
| TSCA | Yes |
| storageNote1 | Hygroscopic |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only