missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium triethylborohydride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
$248.47 - $248.47
Specifications
| Chemical Name or Material | Sodium triethylborohydride |
|---|---|
| Name Note | 1M solution in THF |
| CAS | 17979-81-6,109-99-9 |
| Color | Colorless |
| Density | 0.8900g/mL |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC200031000
|
Thermo Scientific Chemicals
200031000 |
100 mL | AcroSeal™ Glass bottle |
Each for $248.47
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
| Sodium triethylborohydride | |
| 17979-81-6,109-99-9 | |
| 0.8900g/mL | |
| AcroSeal™ Glass bottle | |
| NaB(C2H5)3H | |
| 0.89 | |
| Solubility in water: rzacts. | |
| [Na+].CC[BH-](CC)CC | |
| 121.99 | |
| 121.99 |
| 1M solution in THF | |
| Colorless | |
| −17°C | |
| C6H16BNa | |
| MFCD00011704 | |
| sodium triethylborohydride, sodium triethylborate, sodium triethylhydroborate, sodium triethylhydridoborate, sodium triethylhydroborate 1-, sodium triethylborohydride solution, sodium triethyl-? 2-boranuide, sodiumtriethylborohydride 1m in toluene | |
| YDTZLEUIYNMRLQ-UHFFFAOYSA-N | |
| sodium;triethylboron(1-) | |
| 23667700 | |
| Solution |
RUO – Research Use Only