missing translation for 'onlineSavingsMsg'
Learn More

Sodium tin(IV) oxide trihydrate, 96%

CAS: 12209-98-2 | H6Na2O6Sn | 266.732 g/mol

Supplier:  Thermo Scientific Chemicals A1861136

 View more versions of this product

Catalog No. AAA1861136


Only null left
Explore available promotions

Description

Description

Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.

Solubility
Soluble in water. Insoluble in ethanol and acetone.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Sodium tin(IV) oxide trihydrate
500 g
MFCD00150189
sodium stannate trihydrate, unii-nj7c1v83kg, nj7c1v83kg, sodium stannate inci, sodium stannate vandf, disodium stannate, trihydrate, sodium stannate iv trihydrate mi, 2na.so3n.3h2o, sodiumstannatetrihydrate, sodium tin iv oxide trihydrate
SFXJSNATBHJIDS-UHFFFAOYSA-N
disodium;dioxido(oxo)tin;trihydrate
21977542
98%
12209-98-2
H6Na2O6Sn
14,8677
Soluble in water. Insoluble in ethanol and acetone.
O.O.O.[O-][Sn](=O)[O-].[Na+].[Na+]
266.732
266.75 (212.67 Anhydrous)
Odorless

Chemical Identifiers

12209-98-2
266.732
SFXJSNATBHJIDS-UHFFFAOYSA-N
21977542
O.O.O.[O-][Sn](=O)[O-].[Na+].[Na+]
H6Na2O6Sn
MFCD00150189
sodium stannate trihydrate, unii-nj7c1v83kg, nj7c1v83kg, sodium stannate inci, sodium stannate vandf, disodium stannate, trihydrate, sodium stannate iv trihydrate mi, 2na.so3n.3h2o, sodiumstannatetrihydrate, sodium tin iv oxide trihydrate
disodium;dioxido(oxo)tin;trihydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 235-030-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only