Learn More
Sodium tin(IV) oxide trihydrate, 96%
CAS: 12209-98-2 | H6Na2O6Sn | 266.732 g/mol
Supplier: Thermo Scientific Chemicals A1861122
Description
Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.
Solubility
Soluble in water. Insoluble in ethanol and acetone.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Sodium tin(IV) oxide trihydrate | |
| 100 g | |
| MFCD00150189 | |
| sodium stannate trihydrate, unii-nj7c1v83kg, nj7c1v83kg, sodium stannate inci, sodium stannate vandf, disodium stannate, trihydrate, sodium stannate iv trihydrate mi, 2na.so3n.3h2o, sodiumstannatetrihydrate, sodium tin iv oxide trihydrate | |
| SFXJSNATBHJIDS-UHFFFAOYSA-N | |
| disodium;dioxido(oxo)tin;trihydrate | |
| 21977542 | |
| 98% |
| 12209-98-2 | |
| H6Na2O6Sn | |
| 14,8677 | |
| Soluble in water. Insoluble in ethanol and acetone. | |
| O.O.O.[O-][Sn](=O)[O-].[Na+].[Na+] | |
| 266.732 | |
| 266.75 (212.67 Anhydrous) | |
| Odorless |
Chemical Identifiers
| 12209-98-2 | |
| 266.732 | |
| SFXJSNATBHJIDS-UHFFFAOYSA-N | |
| 21977542 | |
| O.O.O.[O-][Sn](=O)[O-].[Na+].[Na+] |
| H6Na2O6Sn | |
| MFCD00150189 | |
| sodium stannate trihydrate, unii-nj7c1v83kg, nj7c1v83kg, sodium stannate inci, sodium stannate vandf, disodium stannate, trihydrate, sodium stannate iv trihydrate mi, 2na.so3n.3h2o, sodiumstannatetrihydrate, sodium tin iv oxide trihydrate | |
| disodium;dioxido(oxo)tin;trihydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 235-030-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only