missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, Certified, 0.0500N ±0.0005N (0.05M), LabChem™
Description
Sodium Thiosulfate, Certified, 0.0500N ±0.0005N (0.05M)
Specifications
Specifications
| Chemical Name or Material | Sodium Thiosulfate |
| Color | Colorless |
| CAS | 10102-17-7, 7732-18-5, 497-19-8 |
| CAS Max % | 1.24, 98.74, 0.02 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Molecular Formula | H10Na2O8S2 |
| Linear Formula | Na2S2O3·5H2O |
| Synonym | sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate |
| Solubility Information | Soluble in water |
| Show More |