missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiomethoxide, 95%, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94 to 106% (Iodimetry) |
| Packaging | Glass bottle |
| Linear Formula | CH3SNa |
| Quantity | 1 g |
| Synonym | sodium thiomethoxide, sodium methanethiolate, sodium thiomethylate, methyl mercaptan sodium salt, methanethiol sodium salt, sodiumthiomethoxide, methanethiol, sodium salt, sodiummethanethiolate, sodium methyl mercaptide, methylsulfanyl sodium |
| Solubility Information | Solubility in water: freely soluble. |
| IUPAC Name | sodium;methanethiolate |
| Formula Weight | 70.09 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Flammable solid. Toxic if swallowed. Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove conta |
| EINECSNumber | 225-969-9 |
RUO – Research Use Only