missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Sodium Tetrakis(4-Fluorophenyl)Borate Dihydrate
Description
Reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.
Specifications
Specifications
| CAS | 207683-22-5 |
| Molecular Formula | C24H16BF4Na -+ 2H2O |
| Linear Formula | (FC6H4)4BNa-+2H2O |
| MDL Number | MFCD00149598 |
| Quantity | 5 g |
| Synonym | Cesibor; Tetrakis(4-fluorophenyl)boron sodium |
| Molecular Weight (g/mol) | 450.21 g/mol |
| Biological Activity | Respiratory System |