missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetrafluoroborate 95.0+%, TCI America™

Catalog No. O0494500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0494500G 500 g
O049425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0494500G Supplier TCI America Supplier No. O0494500G

Chemical Identifiers

CAS 13755-29-8
Molecular Formula BF4Na
Molecular Weight (g/mol) 109.79
MDL Number MFCD00003515
InChI Key KGJZTOFHXCFQIV-UHFFFAOYSA-N
Synonym sodium tetrafluoroborate, sodium fluoborate, sodium fluoroborate, sodium borofluoride, sodium boron fluoride, apreton r, sodium boron tetrafluoride, sodium fluoroborate nabf4, sodium tetrafluoroborate 1-, sodium tetrafluroborate
PubChem CID 4343483
IUPAC Name sodium tetrafluoroboranuide
SMILES [Na+].F[B-](F)(F)F

Specifications

Melting Point 384°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Formula Weight 109.79
Percent Purity ≥98.0% (T)
Chemical Name or Material Sodium Tetrafluoroborate
EINECSNumber (1)-0053
RTECSNumber ED2975000
TSCA Yes