missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrafluoroborate, 95%

Catalog No. p-7048822
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1156236 500 g
AA11562A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1156236 Supplier Thermo Scientific Chemicals Supplier No. 01156236

Fluorinating agent | CAS: 13755-29-8 | BF4Na | 109.79 g/mol

Fluorinating agentSodium tetrafluoroborate is used in fluxes, metal finishing and in plating circuits. It is used as a starting material in the preparation of ionic liquids as well as boron trifluoride. It acts as an efficient catalyst for the synthesis of 3,4-dihydropyrimidin-2(1H)-ones and thiones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13755-29-8
Molecular Formula BF4Na
Molecular Weight (g/mol) 109.79
MDL Number MFCD00003515
InChI Key KGJZTOFHXCFQIV-UHFFFAOYSA-N
Synonym sodium tetrafluoroborate, sodium fluoborate, sodium fluoroborate, sodium borofluoride, sodium boron fluoride, apreton r, sodium boron tetrafluoride, sodium fluoroborate nabf4, sodium tetrafluoroborate 1-, sodium tetrafluroborate
PubChem CID 4343483
IUPAC Name sodium;tetrafluoroborate
SMILES [Na+].F[B-](F)(F)F

Specifications

Melting Point 384°C (decomposition)
Color White
Physical Form Granular/Lump
Quantity 500 g
Linear Formula NaBF4
Merck Index 14,8617
Solubility Information Soluble in water.
Formula Weight 109.79
Percent Purity 95%
Sensitivity Hygroscopic
Density 2.47 g/mL
Chemical Name or Material Sodium tetrafluoroborate
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 237-340-6
RTECSNumber ED2975000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only