Learn More
Sodium tetracyanoplatinate(II) hydrate, Premion™, 99.95% (metals basis), Pt 48.4% min, Thermo Scientific Chemicals
$338.95 - $338.95
Chemical Identifiers
| CAS | 699012-94-7 |
|---|---|
| Molecular Formula | C4N4Na2Pt |
| Molecular Weight (g/mol) | 345.13 |
| MDL Number | MFCD00798527 |
| InChI Key | WOIFBVMIVVRIIE-UHFFFAOYSA-N |
| Synonym | disodium tetracyanoplatinumdiuide hydrate, sodium tetracyanoplatinate ii hydrate, premion |
| PubChem CID | 16217382 |
| IUPAC Name | disodium tetracyanoplatinumbis(ylium) |
| SMILES | [Na+].[Na+].N#C[Pt++](C#N)(C#N)C#N |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 699012-94-7 | |
| 345.13 | |
| WOIFBVMIVVRIIE-UHFFFAOYSA-N | |
| 16217382 | |
| [Na+].[Na+].N#C[Pt++](C#N)(C#N)C#N |
| C4N4Na2Pt | |
| MFCD00798527 | |
| disodium tetracyanoplatinumdiuide hydrate, sodium tetracyanoplatinate ii hydrate, premion | |
| disodium tetracyanoplatinumbis(ylium) |
Specifications
| (decomposition) | |
| White | |
| 1 g | |
| MFCD00798527 | |
| WOIFBVMIVVRIIE-UHFFFAOYSA-N | |
| disodium tetracyanoplatinumbis(ylium) | |
| 16217382 | |
| 99.95% | |
| Pt 48.4% min. | |
| (metals basis) |
| 699012-94-7 | |
| Crystalline Powder | |
| C4N4Na2Pt | |
| disodium tetracyanoplatinumdiuide hydrate, sodium tetracyanoplatinate ii hydrate, premion | |
| [Na+].[Na+].N#C[Pt++](C#N)(C#N)C#N | |
| 345.13 | |
| 345.14 (Anhydrous) | |
| Premion | |
| Sodium tetracyanoplatinate(II) hydrate |
Safety and Handling
GHS H Statement
H302-H319-H317-H335
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P363-P501c
H302-H317-H319-H335
EINECSNumber : 239-536-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only