missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrabromoaurate(III) hydrate, 99.99% (Metals basis), Thermo Scientific Chemicals, Premion™

Catalog No. AA4042803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4042803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4042803 Supplier Thermo Scientific Chemicals Supplier No. 04042803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium tetrabromoaurate(III) hydrate participates in the fabricated of core/alloy nanoparticles via the layer-by-layer hydrothermal route ans its growth characteristics and optical properties study.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10378-49-1
Molecular Formula AuBr4Na
Molecular Weight (g/mol) 539.57
MDL Number MFCD00274624
InChI Key PMPPMRNISYJZFW-UHFFFAOYSA-J
Synonym sodium tetrabromoaurate iii hydrate, aubr4.na.h2o, gold sodium bromide, naaubr4*xh2o, sodium tetrabromoaurate iii hydrate, premion, sodium tetrabromoaurate 1--water 1/1/1
PubChem CID 71310037
SMILES [Na+].Br[Au-](Br)(Br)Br

Specifications

Physical Form Crystalline
Quantity 1 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water
IUPAC Name sodium;tetrabromogold(1-);hydrate
Formula Weight 539.59 (Anhydrous)
Odor Odorless
Appearance Crystalline powder
Chemical Name or Material Sodium tetrabromoaurate(III) hydrate
Hazard Category H315-H319-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only