missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate, 98%, for analysis, anhydrous

Catalog No. p-4509882
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC206291000 100 g Plastic bottle
AC206295000 500 g Plastic bottle
AC206290025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC206291000 Supplier Thermo Scientific Chemicals Supplier No. 206291000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
Synonym Borax, fused
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Boiling Point 1575.0°C
Melting Point 741.0°C
Color Orange
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Packaging Plastic bottle
Assay Percent Range 98%
Linear Formula Na2B4O7
Fieser 04,461; 15,295
Merck Index 15,8726
Solubility Information Solubility in water: 26g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 201.22
Percent Purity 98%
Grade Analytical
Chemical Name or Material Sodium tetraborate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage fertility.
May damage the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 215-540-4

RUO – Research Use Only