Learn More
Sodium tert-pentyloxide, 95%
CAS: 14593-46-5 | C5H11NaO | 110.132 g/mol
Supplier: Thermo Scientific Chemicals L1511222
Description
Sodium tert-pentyloxide is a effective base for the alkylation of ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium tert-pentyloxide is a effective base for the alkylation of ketones.
Solubility
Reacts with water.
Notes
Air and Moisture Sensitive, store away form air and water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| Sodium tert-pentyloxide | |
| Odorless | |
| MFCD00064245 | |
| UN3206 | |
| Air and moisture sensitive | |
| Reacts with water. | |
| CCC(C)(C)[O-].[Na+] | |
| 110.132 | |
| 110.13 |
| 14593-46-5 | |
| C5H11NaO | |
| 100 g | |
| 3983987 | |
| sodium tert-pentoxide, sodium 2-methylbutan-2-olate, sodium tert-amylate, sodium t-amylate, sodium-t-amylate, unii-51z39py5z8, sodium-tert-pentoxide, sodium tert-amoxide, 2-butanol, 2-methyl-, sodium salt, tert-amyl alcohol sodium salt | |
| CGRKYEALWSRNJS-UHFFFAOYSA-N | |
| sodium;2-methylbutan-2-olate | |
| 2733478 | |
| 95% |
Chemical Identifiers
| 14593-46-5 | |
| 110.132 | |
| CGRKYEALWSRNJS-UHFFFAOYSA-N | |
| 2733478 | |
| CCC(C)(C)[O-].[Na+] |
| C5H11NaO | |
| MFCD00064245 | |
| sodium tert-pentoxide, sodium 2-methylbutan-2-olate, sodium tert-amylate, sodium t-amylate, sodium-t-amylate, unii-51z39py5z8, sodium-tert-pentoxide, sodium tert-amoxide, 2-butanol, 2-methyl-, sodium salt, tert-amyl alcohol sodium salt | |
| sodium;2-methylbutan-2-olate |
Safety and Handling
GHS H Statement
H251-H228-H314-H318
Self-heating: may catch fire.
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P235+P410-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H228-H312+H332-H314-H335-X
DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.
EINECSNumber : 238-369-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only