missing translation for 'onlineSavingsMsg'
Learn More

Sodium tert-Pentoxide 97.0+%, TCI America™

Catalog No. p-7136001
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
S095025G 25 g
S0950100G 100 g
2 options

Catalog No. S095025G

Supplier: TCI America S095025G

Only null left

Chemical Identifiers

CAS 14593-46-5
Molecular Formula C5H11NaO
Molecular Weight (g/mol) 110.132
MDL Number MFCD00064245
InChI Key CGRKYEALWSRNJS-UHFFFAOYSA-N
Synonym sodium tert-pentoxide, sodium 2-methylbutan-2-olate, sodium tert-amylate, sodium t-amylate, sodium-t-amylate, unii-51z39py5z8, sodium-tert-pentoxide, sodium tert-amoxide, 2-butanol, 2-methyl-, sodium salt, tert-amyl alcohol sodium salt
PubChem CID 2733478
IUPAC Name sodium;2-methylbutan-2-olate
SMILES CCC(C)(C)[O-].[Na+]

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3206
Formula Weight 110.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium tert-Pentoxide
EINECSNumber (2)-0206
TSCA Yes