missing translation for 'onlineSavingsMsg'
Learn More

Sodium tert-pentoxide, 30% in 2-MeTHF, AcroSeal™, Thermo Scientific™

$165.12 - $898.89

Specifications

CAS 14593-46-5 , 96-47-9
Molecular Formula C5H11NaO
Molecular Weight (g/mol) 110.13
InChI Key CGRKYEALWSRNJS-UHFFFAOYSA-N
Synonym sodium tert-pentoxide, sodium 2-methylbutan-2-olate, sodium tert-amylate, sodium t-amylate, sodium-t-amylate, unii-51z39py5z8, sodium-tert-pentoxide, sodium tert-amoxide, 2-butanol, 2-methyl-, sodium salt, tert-amyl alcohol sodium salt
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC252531000
View Documents Promotion Details
N/A 100mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC252538000
View Documents Promotion Details
N/A 800mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

14593-46-5 , 96-47-9
110.13
sodium tert-pentoxide, sodium 2-methylbutan-2-olate, sodium tert-amylate, sodium t-amylate, sodium-t-amylate, unii-51z39py5z8, sodium-tert-pentoxide, sodium tert-amoxide, 2-butanol, 2-methyl-, sodium salt, tert-amyl alcohol sodium salt
CCC(C)(C)[O-].[Na+]
C5H11NaO
CGRKYEALWSRNJS-UHFFFAOYSA-N
2733478
sodium;2-methylbutan-2-olate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.
May form explosive peroxides.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 238-639-4

SDS
Documents

Documents

RUO – Research Use Only