missing translation for 'onlineSavingsMsg'
Learn More

Sodium tellurite(IV), 99.5% (metals basis)

Catalog No. AA4177818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4177818 50 g
AA4177809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4177818 Supplier Thermo Scientific Chemicals Supplier No. 04177818

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10102-20-2 | H10Na2O9Te | 327.65 g/mol

Sodium tellurite(IV) acts as an intermediate used in the extraction of the tellurium. It is used to make corrosion-resistant electroplated nickel. It is also used to add a bluish coating to iron, steel and aluminum. It serves as a growth medium for bacteria in microbiology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10102-20-2
Molecular Formula H10Na2O9Te
Molecular Weight (g/mol) 327.65
MDL Number MFCD00036141
InChI Key OVBVNEXDNNHXIU-UHFFFAOYSA-L
Synonym sodium tellurite, disodium tellurite, sodium tellurate iv, disodium trioxotellurate, unii-0bb57la23y, tellurous acid, disodium salt, sodium tellurite iv, telluric acid h2teo3 , disodium salt, telluric acid, disodium salt
PubChem CID 24935
IUPAC Name disodium;tellurite
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Te]([O-])(=O)=O

Specifications

Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Soluble in water.
Formula Weight 221.58
Appearance White powder
Chemical Name or Material Sodium tellurite(IV)
Hazard Category H301
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 233-268-4
RTECSNumber WY2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only