missing translation for 'onlineSavingsMsg'
Learn More

Sodium tellurite(IV), 97%

CAS: 10102-20-2 | H10Na2O9Te | 327.65 g/mol

$368.95 - $1361.05

Chemical Identifiers

CAS 10102-20-2
Molecular Formula H10Na2O9Te
Molecular Weight (g/mol) 327.65
MDL Number MFCD00036141
InChI Key OVBVNEXDNNHXIU-UHFFFAOYSA-L
Synonym sodium tellurite, disodium tellurite, sodium tellurate iv, disodium trioxotellurate, unii-0bb57la23y, tellurous acid, disodium salt, sodium tellurite iv, telluric acid h2teo3 , disodium salt, telluric acid, disodium salt
PubChem CID 24935
IUPAC Name disodium;tellurite
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Te]([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1563318
View Documents
Thermo Scientific Chemicals
A1563318
50 g
Each for $368.95
Only null left
 
AAA1563330
View Documents
Thermo Scientific Chemicals
A1563330
250 g
Each for $1,361.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10102-20-2
327.65
OVBVNEXDNNHXIU-UHFFFAOYSA-L
24935
O.O.O.O.O.[Na+].[Na+].[O-][Te]([O-])(=O)=O
H10Na2O9Te
MFCD00036141
sodium tellurite, disodium tellurite, sodium tellurate iv, disodium trioxotellurate, unii-0bb57la23y, tellurous acid, disodium salt, sodium tellurite iv, telluric acid h2teo3 , disodium salt, telluric acid, disodium salt
disodium;tellurite

Specifications

10102-20-2
H10Na2O9Te
MFCD00036141
14,8685
OVBVNEXDNNHXIU-UHFFFAOYSA-L
disodium;tellurite
24935
97%
50 g
Na2TeO3
UN3284
sodium tellurite, disodium tellurite, sodium tellurate iv, disodium trioxotellurate, unii-0bb57la23y, tellurous acid, disodium salt, sodium tellurite iv, telluric acid h2teo3 , disodium salt, telluric acid, disodium salt
O.O.O.O.O.[Na+].[Na+].[O-][Te]([O-])(=O)=O
327.65
221.58
Sodium tellurite(IV)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TELLURIUM COMPOUND, N.O.S.

EINECSNumber : 233-268-4

RTECSNumber : WY2450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only