missing translation for 'onlineSavingsMsg'
Learn More

Sodium tellurite(IV), 97%

Catalog No. AAA1563318
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1563318 50 g
AAA1563330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1563318 Supplier Thermo Scientific Chemicals Supplier No. A1563318

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10102-20-2 | H10Na2O9Te | 327.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10102-20-2
Molecular Formula H10Na2O9Te
Molecular Weight (g/mol) 327.65
MDL Number MFCD00036141
InChI Key OVBVNEXDNNHXIU-UHFFFAOYSA-L
Synonym sodium tellurite, disodium tellurite, sodium tellurate iv, disodium trioxotellurate, unii-0bb57la23y, tellurous acid, disodium salt, sodium tellurite iv, telluric acid h2teo3 , disodium salt, telluric acid, disodium salt
PubChem CID 24935
IUPAC Name disodium;tellurite
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Te]([O-])(=O)=O

Specifications

Quantity 50 g
Linear Formula Na2TeO3
UN Number UN3284
Merck Index 14,8685
Formula Weight 221.58
Percent Purity 97%
Chemical Name or Material Sodium tellurite(IV)
Hazard Category H301
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TELLURIUM COMPOUND, N.O.S.
EINECSNumber 233-268-4
RTECSNumber WY2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only