missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tartrate, Dihydrate, Purified, Spectrum™ Chemical

Catalog No. p-7158289
Change view
Click to view available options
Quantity:
500 g
2.5 kg
100 lb.
Packaging:
Fiber Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606875 500 g Poly Bottle
18606877 100 lb. Fiber Drum
18606876 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606875 Supplier Spectrum Chemical Mfg Cor Supplier No. S1488500GM
Only null left

S1488, 6106-24-7, Na2C4H4O6·2H2O

Spectrum™ Chemical Sodium Tartrate, Dihydrate, Purified , also known as Sal tartar or Disodium tartrate, is used as a binding agent in certain foods and as an emulsifier. The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
IUPAC Name disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

CAS Max % 1
pH Approx. 9
Packaging Poly Bottle
Quantity 500 g
Grade Purified