missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tartrate Dihydrate, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCS0890500
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCS0890500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCS0890500 Supplier Ricca Chemical Company Supplier No. RDCS0890500B1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
Synonym unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
PubChem CID 131855972
IUPAC Name disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Percent Purity 99 to 101%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Sodium Tartrate, Dihydrate
ShelfLife 60 months