missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfide Nonahydrate, Honeywell Fluka™
ACS Reagent,≥98.0%
$178.92 - $602.55
Chemical Identifiers
| CAS | 1313-84-4 |
|---|---|
| Molecular Formula | H18Na2O9S |
| Molecular Weight (g/mol) | 240.17 |
| MDL Number | MFCD00149184 |
| InChI Key | ZGHLCBJZQLNUAZ-UHFFFAOYSA-N |
| IUPAC Name | disodium nonahydrate sulfanediide |
| SMILES | O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 1313-84-4 | |
| 240.17 | |
| ZGHLCBJZQLNUAZ-UHFFFAOYSA-N | |
| O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] |
| H18Na2O9S | |
| MFCD00149184 | |
| disodium nonahydrate sulfanediide |
Specifications
| Colorless or Yellow | |
| 100 g | |
| Na2S·9H2O | |
| 1849 | |
| O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] | |
| 240.17 | |
| ≥98% | |
| Sodium Sulfide Nonahydrate |
| 1313-84-4 | |
| H18Na2O9S | |
| MFCD00149184 | |
| ZGHLCBJZQLNUAZ-UHFFFAOYSA-N | |
| disodium nonahydrate sulfanediide | |
| 240.18g/mol | |
| ACS Reagent |
Safety and Handling
P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P391
H302-H311-H314-H400
EINECSNumber : 215-211-5
RTECSNumber : WE1925000