missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfide hydrate, tech., Thermo Scientific Chemicals

Catalog No. AA1166436
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1166436 500 g
AA11664A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1166436 Supplier Thermo Scientific Chemicals Supplier No. 01166436

Sodium sulfide hydrate is used as an oxygen scavenger in water treatment, bleaching agent in textile industry, desulfurising and as a dechlorinating agent and in leather industry, sulfonation and sulfomethylation agent in chemical industry. It is involved in the production of rubber chemicals and sulfur dyes. It is also used to prepare 3-nitroaniline from 1,3-dinitrobenzene and thioethers from alkyl halide. It plays an important role in other applications such as ore flotation, oil recovery, making dyes, and detergent. In addition, it is used in pulp and paper industry in the Kraft process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27610-45-3
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00149183
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
PubChem CID 45051681
SMILES [Na+].[Na+].[S--]

Specifications

Melting Point ∽50°C (decomposition)
Physical Form Flakes
Quantity 500 g
UN Number UN1849
Merck Index 14,8681
Solubility Information Soluble in water; Slightly soluble in alcohol; Insoluble in etherSoluble in water; Slightly soluble in alcohol; Insoluble in ether.
IUPAC Name disodium sulfanediide
Formula Weight 78.04 (anhydrous)
Grade Technical
Sensitivity Hygroscopic; Light sensitive
Density 1.427 g/mL
Chemical Name or Material Sodium sulfide hydrate
Show More Show Less
Hazard Category H290-H302-H311-H314
Hazard Statement GHS H Statement
H311-H314-H318-H302
Precautionary Statement P234-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM SULFIDE, HYDRATED
EINECSNumber 215-211-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only