missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfide, Fused Flakes, Technical, Reagents
Sodium Sulfide, Fused Flakes, Technical, Technical, for general laboratory use. Manufactured in ISO 9001 facility.
$130.97 - $1512.70
Chemical Identifiers
| CAS | 27610-45-3 |
|---|---|
| Molecular Formula | Na2S |
| Molecular Weight (g/mol) | 78.04 |
| MDL Number | MFCD00149183 |
| InChI Key | GRVFOGOEDUUMBP-UHFFFAOYSA-N |
| Synonym | Sodium sulphide scales |
| IUPAC Name | disodium sulfanediide |
| SMILES | [Na+].[Na+].[S--] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-336-959
|
Labchem Inc
C232600012F4 |
12 kg |
Each for $1,512.70
|
|
|||||
|
01-336-958
|
Labchem Inc
C232600025A5 |
2.5 kg |
Each for $399.13
|
|
|||||
|
01-336-957
|
Labchem Inc
C2326000500A5 |
500 g |
Each for $130.97
|
|
|||||
Chemical Identifiers
| 27610-45-3 | |
| 78.04 | |
| GRVFOGOEDUUMBP-UHFFFAOYSA-N | |
| disodium sulfanediide |
| Na2S | |
| MFCD00149183 | |
| Sodium sulphide scales | |
| [Na+].[Na+].[S--] |
Specifications
| 50°C | |
| 27610-45-3 | |
| 12 kg | |
| MFCD00149183 | |
| GRVFOGOEDUUMBP-UHFFFAOYSA-N | |
| disodium sulfanediide | |
| Technical | |
| 1.856 |
| Colorless to Slight Yellow | |
| Solid | |
| Na2S | |
| Sodium sulphide scales | |
| [Na+].[Na+].[S--] | |
| 78.04 | |
| Fiber Drum | |
| Sodium Sulfide, Fused Flakes, Technical |