missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfide, Fused Flakes, Technical, Reagents

Catalog No. 01336959
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336959 12 kg
01336958 2.5 kg
01336957 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336959 Supplier Labchem Inc Supplier No. C232600012F4

Sodium Sulfide, Fused Flakes, Technical, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 27610-45-3
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00149183
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
Synonym Sodium sulphide scales
IUPAC Name disodium sulfanediide
SMILES [Na+].[Na+].[S--]

Specifications

Melting Point 50°C
Color Colorless to Slight Yellow
Physical Form Solid
Quantity 12 kg
Grade Technical
Packaging Fiber Drum
Specific Gravity 1.856
Chemical Name or Material Sodium Sulfide, Fused Flakes, Technical