missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfide, Fused Flakes, Approx. 60%, Spectrum™ Chemical

Catalog No. 18606845
Change view
Click to view available options
Quantity:
45 kg
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606845 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606845 Supplier Spectrum Chemical Mfg Cor Supplier No. S147045KGBL

S1470, 27610-45-3, Na2S·xH2O

Spectrum™ Chemical Sodium Sulfide, Fused Flakes , also known as disodium sulfide, is used as an oxygen scavenger agent in water treatment as well as a metals precipitant. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 27610-45-3
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00149183
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
IUPAC Name disodium sulfanediide
SMILES [Na+].[Na+].[S--]

Specifications

CAS Max % 1
Quantity 45 kg
Percent Purity ∼60%
Grade Reagent
Packaging Fiber Drum