missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfide, Fused Flakes, Approx. 60%, Spectrum™ Chemical
S1470, 27610-45-3, Na2S·xH2O
$387.07 - $14447.60
Chemical Identifiers
| CAS | 27610-45-3 |
|---|---|
| Molecular Formula | Na2S |
| Molecular Weight (g/mol) | 78.04 |
| MDL Number | MFCD00149183 |
| InChI Key | GRVFOGOEDUUMBP-UHFFFAOYSA-N |
| IUPAC Name | disodium sulfanediide |
| SMILES | [Na+].[Na+].[S--] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606842
|
Spectrum Chemical Mfg Cor
S1470500GM |
500 g | Amber Glass Bottle |
Each for $387.07
|
|
||||
|
18606846
|
Spectrum Chemical Mfg Cor
S1470400LBBL |
400 lb. | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18606843
|
Spectrum Chemical Mfg Cor
S147025KG |
2.5 kg | Poly Pail |
Each for $1,274.03
|
|
||||
|
18606844
|
Spectrum Chemical Mfg Cor
S147012KG |
12 kg | Poly Pail |
Each for $2,708.13
|
|
||||
Description
Spectrum™ Chemical Sodium Sulfide, Fused Flakes , also known as disodium sulfide, is used as an oxygen scavenger agent in water treatment as well as a metals precipitant. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 27610-45-3 | |
| 78.04 | |
| GRVFOGOEDUUMBP-UHFFFAOYSA-N | |
| [Na+].[Na+].[S--] |
| Na2S | |
| MFCD00149183 | |
| disodium sulfanediide |
Specifications
| 27610-45-3 | |
| 500 g | |
| MFCD00149183 | |
| [Na+].[Na+].[S--] | |
| 78.04 | |
| Reagent |
| 1 | |
| Na2S | |
| GRVFOGOEDUUMBP-UHFFFAOYSA-N | |
| disodium sulfanediide | |
| ∼60% | |
| Amber Glass Bottle |