missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfathiazole, Sesquihydrate, 99%, Spectrum™ Chemical
S1462, C9H8N3NaO2S2·1.5H2O
$1952.03 - $3342.53
Specifications
| CAS Max % | 1 |
|---|---|
| Quantity | 500 g |
| Percent Purity | 99% |
| Grade | Reagent |
| Loss on Drying | 0.09 |
| Packaging | Poly Bottle |
Description
Spectrum™ Chemical Sodium Sulfathiazole, Sesquihydrate , an organosulfur compound, can be used as a short-acting sulfa drug and as an oral and topical antimicrobial, Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Specifications
| 1 | |
| 99% | |
| 0.09 |
| 500 g | |
| Reagent | |
| Poly Bottle |