missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™

Catalog No. 6001995
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001995 1 kg
6001996 2.5 kg
60044811 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001995 Supplier Honeywell Chemicals Supplier No. 134641KG
Only null left

Puriss., meets analytical specification of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Physical Form Anhydrous
Quantity 1 kg
Linear Formula Na2SO4
UN Number NONH for all modes of transport
IUPAC Name disodium;sulfate
Formula Weight 142.04g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Density 2.68g/mL at 25°C
Chemical Name or Material Sodium Sulfate, Meets analytical specification of Ph. Eur.Bp, USP
EINECSNumber 231-820-9
RTECSNumber WE1650000