missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate decahydrate, extra pure

CAS: 7727-73-3 | Na2O410H2O | 322.2 g/mol

$67.12 - $139.47

Chemical Identifiers

CAS 7727-73-3
Molecular Formula Na2O410H2O
Molecular Weight (g/mol) 322.2
MDL Number MFCD00149189
InChI Key RSIJVJUOQBWMIM-UHFFFAOYSA-L
Synonym sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
PubChem CID 62649
ChEBI CHEBI:32586
IUPAC Name disodium;sulfate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC125012500
SDS View Documents
Thermo Scientific Chemicals
125012500
250 g Plastic bottle
Each for $67.12
Only null left
 
AC125010010
SDS View Documents
Thermo Scientific Chemicals
125010010
1 kg Plastic bottle
Each for $139.47
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7727-73-3
322.2
RSIJVJUOQBWMIM-UHFFFAOYSA-L
62649
disodium;sulfate;decahydrate
Na2O410H2O
MFCD00149189
sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
CHEBI:32586
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]

Specifications

884.0°C
7727-73-3
Crystalline Powder and/or Crystals
Na2O410H2O
MFCD00149189
14,868
Solubility in water: soluble
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]
322.2
CHEBI:32586
≥99.0%
Plastic bottle
White
5.2 to 9.2 (5% soln.)
250 g
Na2SO4·10H2O
01,1103
sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
RSIJVJUOQBWMIM-UHFFFAOYSA-L
disodium;sulfate;decahydrate
62649
322.2
Extra Pure
Sodium sulfate decahydrate
Videos
SDS
Documents

Documents

RUO – Research Use Only