missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate decahydrate, 99%

Catalog No. p-7022789
Change view
Click to view available options
Quantity:
25,000 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA157020B 1000 g
AAA157020I 5000 g
AAA157020G 25,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA157020B Supplier Thermo Scientific Chemicals Supplier No. A157020B
Only null left

CAS: 7727-73-3 | H20Na2O14S | 322.186 g/mol

Sodium sulfate decahydrate is used as a laboratory reagent to precipitate protein. It is largely used as filler in powdered home laundry detergents. It facilitates in leveling, reducing negative charges on fibers, thereby allows dye to penetrate evenly. It avoids the corrosion of stainless steel vessels used in dyeing. It is also used as a laxative and effectively replaces the drug paracetamol in the body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium sulfate decahydrate is used as a laboratory reagent to precipitate protein. It is largely used as filler in powdered home laundry detergents. It facilitates in leveling, reducing negative charges on fibers, thereby allows dye to penetrate evenly. It avoids the corrosion of stainless steel vessels used in dyeing. It is also used as a laxative and effectively replaces the drug paracetamol in the body.

Solubility
Soluble in water and glycerin. Insoluble in ethanol and methanol.

Notes
Stable. Incompatible with strong acids, aluminum and magnesium.

Chemical Identifiers

CAS 7727-73-3
Molecular Formula H20Na2O14S
Molecular Weight (g/mol) 322.186
MDL Number MFCD00149189
InChI Key RSIJVJUOQBWMIM-UHFFFAOYSA-L
Synonym sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
PubChem CID 62649
ChEBI CHEBI:32586
IUPAC Name disodium;sulfate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point ∼32°C
pH 5.2 to 8.0
Quantity 1000 g
Merck Index 14,8680
Solubility Information Soluble in water and glycerin. Insoluble in ethanol and methanol.
Formula Weight 322.19 (142.04 Anhydrous)
Percent Purity 99%
Odor Odorless
Density 1.46
Chemical Name or Material Sodium sulfate decahydrate
EINECSNumber 231-820-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only